missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylmagnesium bromide, 0.8M solution in THF, AcroSeal™

Product Code. p-2799185 Shop All Thermo Scientific Chemicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
missing translation for 'unitSize'
100mL
800mL
This item is not returnable. View return policy

Product Code. 10240703

missing translation for 'mfr': Thermo Scientific Acros 377231000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 352-13-6 | C6H4BrFMg | 199.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-13-6, 109-99-9
Molecular Formula C6H4BrFMg
Molecular Weight (g/mol) 199.31
MDL Number MFCD00009667
InChI Key QYBFFRXNNFXREA-UHFFFAOYSA-M
Synonym 4-fluorophenylmagnesium bromide, magnesium, bromo 4-fluorophenyl, bromo 4-fluorophenyl magnesium, p-fluorophenylmagnesium bromide, 4-fluorophenyl magnesium bromide, 4-f-c6h4-mgbr, 4-fluorophenylmagnesiumbromide, 4-fluorphenylmagnesium bromide, 4-flourophenylmagnesium bromide, 4-fluorophenyl magnesiumbromide, grignard reagent
PubChem CID 2734897
IUPAC Name bromo(4-fluorophenyl)magnesium
SMILES FC1=CC=C([Mg]Br)C=C1

Specifications

Color Yellow to Brown
Melting Point 5.0°C
Linear Formula F(C6H6)MgBr
Density 1.0100g/mL
Solubility Information Solubility in water: vigorous reaction.
Formula Weight 199.3
Percent Purity 14 to 17%
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.01
Quantity 100 mL
Flash Point −21°C
Physical Form Solution
Chemical Name or Material 4-Fluorophenylmagnesium bromide, 0.8M Solution in THF
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Fluorophenylmagnesium bromide, 0.8M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H261-In contact with water releases flammable gases.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.