missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10402242 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal Glass Bottle in Tin Can
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10402242

Brand: Thermo Scientific Acros 377371000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-13-6, 60-29-7
Molecular Formula C6H4BrFMg
Molecular Weight (g/mol) 199.31
MDL Number MFCD00009667
InChI Key QYBFFRXNNFXREA-UHFFFAOYSA-M
Synonym 4-fluorophenylmagnesium bromide, magnesium, bromo 4-fluorophenyl, bromo 4-fluorophenyl magnesium, p-fluorophenylmagnesium bromide, 4-fluorophenyl magnesium bromide, 4-f-c6h4-mgbr, 4-fluorophenylmagnesiumbromide, 4-fluorphenylmagnesium bromide, 4-flourophenylmagnesium bromide, 4-fluorophenyl magnesiumbromide, grignard reagent
PubChem CID 2734897
SMILES FC1=CC=C([Mg]Br)C=C1

Specifications

Color Undesignated
CAS Min % 38.22, 57.96
CAS Max % 42.04, 61.78
Linear Formula F(C6H4)MgBr
Concentration 2 to 2.2M
Density 1.0430g/mL
IUPAC Name bromo(4-fluorophenyl)magnesium
Formula Weight 199.3
Packaging AcroSeal Glass Bottle in Tin Can
Quantity 100 mL
Physical Form Solution
Chemical Name or Material 4-Fluorophenylmagnesium bromide, 2M Solution in Diethyl Ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.