missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophthalic anhydride, 98%, Thermo Scientific Chemicals

Product Code. 11449287
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Fluorophthalic anhydride is used in the preparation of 5-fluoro-2-pyridin-3-ylmethyl-isoindole-1,3-dione by reacting with C-pyridin-3-yl-methylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophthalic anhydride is used in the preparation of 5-fluoro-2-pyridin-3-ylmethyl-isoindole-1,3-dione by reacting with C-pyridin-3-yl-methylamine.

Solubility
Soluble in sodium hydroxide.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 319-03-9
Molecular Formula C8H3FO3
Molecular Weight (g/mol) 166.11
MDL Number MFCD00191363
InChI Key XVMKZAAFVWXIII-UHFFFAOYSA-N
Synonym 4-fluorophthalic anhydride, 5-fluoroisobenzofuran-1,3-dione, 5-fluoro-1,3-isobenzofurandione, 1,3-isobenzofurandione, 5-fluoro, 4-fluorophthalicanhydride, 4-fluorophthalic acid anhydride, 5-fluoro-1,3-dihydro-2-benzofuran-1,3-dione, 4-fluorobenzene-1,2-dicarboxylic anhydride, 5-fluoro-isobenzofuran-1,3-dione
PubChem CID 67572
IUPAC Name 5-fluoro-2-benzofuran-1,3-dione
SMILES FC1=CC=C2C(=O)OC(=O)C2=C1

Specifications

Melting Point 74°C to 79°C
Density 1.5
Boiling Point 147°C to 149°C (20 mmHg)
Quantity 5 g
Beilstein 131281
Sensitivity Moisture sensitive
Solubility Information Soluble in sodium hydroxide.
Formula Weight 166.11
Percent Purity 98%
Chemical Name or Material 4-Fluorophthalic anhydride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'