missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorothioanisole, 97%, Thermo Scientific™

Product Code. 11331787
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-15-3
Molecular Formula C7H7FS
Molecular Weight (g/mol) 142.191
MDL Number MFCD00040829
InChI Key XFUMHENRNCUHOH-UHFFFAOYSA-N
Synonym 4-fluorothioanisole, 1-fluoro-4-methylthio benzene, 4-fluorophenyl methyl sulfide, 4-fluoro thioanisole, 1-fluoro-4-methylsulfanyl-benzene, 1-fluoro-4-methylsulfanyl benzene, p-fluoro methylthio benzene, benzene, 1-fluoro-4-methylthio, 4-fluoro-1-methylthiobenzene, pubchem10641
PubChem CID 3083565
IUPAC Name 1-fluoro-4-methylsulfanylbenzene
SMILES CSC1=CC=C(C=C1)F

Specifications

Density 1.167
Boiling Point 89°C to 90°C (31 mmHg)
Flash Point 73°C (163°F)
Odor Stench
Refractive Index 1.55
Quantity 5 g
UN Number UN3334
Beilstein 2041508
Formula Weight 142.2
Percent Purity 97%
Chemical Name or Material 4-Fluorothioanisole
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'