missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Formylnaphthalene-1-boronic acid, 95%

Product Code. 15439907
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 332398-52-4
Melting Point 230°C
Molecular Formula C11H9BO3
MDL Number MFCD01632205
Quantity 250 mg
Synonym 4-formylnaphthalen-1-yl boronic acid, 4-formylnaphthalene-1-boronic acid, 4-formyl-1-naphthalene boronic acid, boronic acid, 4-formyl-1-naphthalenyl, boronic acid, b-4-formyl-1-naphthalenyl, acmc-209hzm, 4-formyl-1-naphthylboronic acid, 4-formyl-1-naphthaleneboronic acid, 4-formyl-1-naphtalene boronic acid, 4-formyl-1-naphthalenyl boronic acid
InChI Key KBHGTTWVFRLPRO-UHFFFAOYSA-N
SMILES OB(O)C1=C2C=CC=CC2=C(C=O)C=C1
IUPAC Name (4-formylnaphthalen-1-yl)boronic acid
Molecular Weight (g/mol) 200.00
PubChem CID 2773424
Formula Weight 200
Percent Purity 95%
Chemical Name or Material 4-Formylnaphthalene-1-boronic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.