missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ 4 ft. RXPert Double Filtered System

RXPert Double Filtered Balance Systems are Class I biological safety cabinets that contain airborne particulates and powders, providing user protection during hazardous drug compounding or pharmaceutical manipulation.

Brand:  Labconco™ 3971483


Additional Details : Weight : 64.00000kg

Product Code. 16804463

  • £13705.00 / Each
Estimated Shipment: 29-10-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

All Models Feature

  • USP compliance
  • Patented design
  • Deep interior of 23.4 in. (59.4 cm) or 30.0 in. (76.2 cm) depending on model
  • Ergonomic air foil with aerodynamic Clean-Sweep™ airflow openings
  • Upper dilution air supply
  • Upper containment sash foil
  • Zone-perforated, pivoting rear baffle
  • Side-entry air foils
  • Exhaust HEPA filter, 99.99% efficient on particles 0.3 micron in size
  • True bag-in/bag-out filter disposal system

Exclusive Features of RXPert Double Filtered Balance Systems

  • True bag-in/bag-out HEPA filter disposal system allows for safe filter replacement.
  • Optimal HEPA filter location permits leak-detection scan of entire surface.
  • Upper dilution air supply sweeps clean air to the back of sash to keep contaminants away from the operator.
  • Upper containment sash foil directs contaminants away from the user’s breathing zone.
  • Patented Clean-Sweep™ air foil allows air to sweep the work surface to maximize containment.
  • Patented rear baffle with zones of perforations promotes horizontal laminar airflow.
  • The baffle pivots down for cleaning.
  • Deep 23.4 in. or 30.0 in. (depending on model) interior easily accommodates large analytical balances.
Specifications

Specifications

Balance Enclosure
Fluorescent
29 in.
49.5 in.
125.7 cm
50/60 Hz
230 V
181.9 kg
121.9 cm
HEPA
ANSI Z9.5, CE, Modified ASHRAE 110, SEFA 1, USP <800> non-sterile compounding
73.7 cm
9.4 in.
23.9 cm
Guardian
401 lb.
48 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers