missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva 4-Gel Caster Complete for MiniVE Vertical Electrophoresis System
Maintains uniform gel temperature with the tank-style lower buffer chamber
Brand: Cytiva 80614612
Code : GE
Additional Details : Weight : 1,468.00000kg
Includes:
- 4 x Notched alumina plate, 10 x 10.5cm
- 5 x Rectangular glass plate, 10 x 10.5cm
- 4 x Clamp
- 100 x Wax paper sheet
- 1 x Space-saver plate
- 5 x Filler sheet
- 1 x Filler plugs set
- 1 x Spacer-Mate assembly template
Description
- A full tank stabilizes the temperature so that lanes are straight and bands are flat
- Gel module is simultaneously a casting stand and an upper buffer chamber with integrated clamps
- Assemble the gel sandwich of plates and spacers, clamp it onto the upper buffer chamber, then pivot the sealing gasket and cast the gel
- Can produce up to four identical gels at one time
- Can be used with Mini-Vertical Unit SE 260 also
- Spacer Thickness: 0.75mm, 1.0mm and 1.5mm
- Number of Gels: 2 - 4
- Gel Capacity: 2-4 Gels
Specifications
MiniVE™ Vertical Electrophoresis System and Mini Vertical unit SE 260 | |
Gel Caster | |
4 Notched Alumina plate,10 x 10.5cm, 5 rectangular Glass plate, 10 x 10.5cm, 4 clamp, 100 Wax Paper sheet, 5 filler sheet, 1 filler plugs set, 1 spacer mate assembly template |