missing translation for 'onlineSavingsMsg'
Learn More

4-Heptanol, 93%, Thermo Scientific Chemicals

Product Code. 10716201
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 589-55-9
CAS Min % 91.0
CAS Max % 100.0
Melting Point -41.5°C
Color Colorless to Yellow
Density 0.8100g/mL
Boiling Point 156°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 91% min. (GC)
Packaging Glass bottle
Molecular Formula C7H16O
Linear Formula CH3(CH2)2CH(OH)C3H7
Refractive Index 1.4175 to 1.4215
MDL Number MFCD00021934
Quantity 5 mL
Specific Gravity 0.81
Synonym 4-heptanol, dipropylcarbinol, unii-yg7b8091bp, di-n-propylcarbinol, heptanol-4, 4-heptyl alcohol, n-heptan-4-ol, acmc-209m8k, yvbculsizwmtfy-uhfffaoysa, ch3 ch2 2choh ch2 2ch3
Solubility Information Solubility in water: immiscible
InChI Key YVBCULSIZWMTFY-UHFFFAOYSA-N
SMILES CCCC(CCC)O
IUPAC Name heptan-4-ol
Molecular Weight (g/mol) 116.2
PubChem CID 11513
Formula Weight 116.2
Percent Purity 93%
Physical Form Liquid
Chemical Name or Material 4-Heptanol, 93%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Heptanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'