missing translation for 'onlineSavingsMsg'
Learn More

4-Hexyloxybenzaldehyde, 99%, Thermo Scientific™

Product Code. 10526982
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 5736-94-7
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.28
MDL Number MFCD00016615
InChI Key GWXUVWKBVROFDM-UHFFFAOYSA-N
Synonym 4-hexyloxy benzaldehyde, 4-hexyloxybenzaldehyde, 4-n-hexyloxybenzaldehyde, p-hexyloxybenzaldehyde, benzaldehyde, 4-hexyloxy, benzaldehyde, p-hexyloxy, p-hexyloxy benzaldehyde, p-hexoxybenzaldehyde, 4-hexyloxbenzaldehyde, p-n-hexyloxybenzaldehyde
PubChem CID 79816
IUPAC Name 4-hexoxybenzaldehyde
SMILES CCCCCCOC1=CC=C(C=C1)C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9920g/mL
Boiling Point 166°C (12.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5285 to 1.5325
Quantity 25 g
Specific Gravity 0.992
Formula Weight 206.28
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Hexyloxybenzaldehyde, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Hexyloxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'