Learn More
4-Hydrazinobenzoic acid, 97%, Thermo Scientific Chemicals
£28.90 - £402.00
Chemical Identifiers
CAS | 619-67-0 |
---|---|
Molecular Formula | C7H8N2O2 |
Molecular Weight (g/mol) | 152.153 |
MDL Number | MFCD00007581 |
InChI Key | PCNFLKVWBDNNOW-UHFFFAOYSA-N |
Synonym | 4-hydrazinobenzoic acid, p-hydrazinobenzoic acid, benzoic acid, 4-hydrazino, p-carboxyphenylhydrazine, 4-carboxyphenyl hydrazine, 4-carboxyphenylhydrazine, 4-hydrazino benzoic acid, benzoic acid, p-hydrazino, unii-b1r368je7o, ccris 7911 |
PubChem CID | 12089 |
IUPAC Name | 4-hydrazinylbenzoic acid |
SMILES | C1=CC(=CC=C1C(=O)O)NN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11421558
|
Thermo Scientific Alfa Aesar
A13874.09 |
10 g |
£28.90
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11431558
|
Thermo Scientific Alfa Aesar
A13874.18 |
50 g |
£93.10
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11441558
|
Thermo Scientific Alfa Aesar
A13874.30 |
250 g |
£402.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
619-67-0 | |
152.153 | |
PCNFLKVWBDNNOW-UHFFFAOYSA-N | |
12089 | |
C1=CC(=CC=C1C(=O)O)NN |
C7H8N2O2 | |
MFCD00007581 | |
4-hydrazinobenzoic acid, p-hydrazinobenzoic acid, benzoic acid, 4-hydrazino, p-carboxyphenylhydrazine, 4-carboxyphenyl hydrazine, 4-carboxyphenylhydrazine, 4-hydrazino benzoic acid, benzoic acid, p-hydrazino, unii-b1r368je7o, ccris 7911 | |
4-hydrazinylbenzoic acid |
Specifications
619-67-0 | |
C7H8N2O2 | |
387378 | |
PCNFLKVWBDNNOW-UHFFFAOYSA-N | |
4-hydrazinylbenzoic acid | |
12089 | |
97% |
∼216°C (decomposition) | |
MFCD00007581 | |
4-hydrazinobenzoic acid, p-hydrazinobenzoic acid, benzoic acid, 4-hydrazino, p-carboxyphenylhydrazine, 4-carboxyphenyl hydrazine, 4-carboxyphenylhydrazine, 4-hydrazino benzoic acid, benzoic acid, p-hydrazino, unii-b1r368je7o, ccris 7911 | |
C1=CC(=CC=C1C(=O)O)NN | |
152.153 | |
152.15 | |
4-Hydrazinobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 210-609-5
missing translation for 'rtecsNumber' : DH1700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only