missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzoic acid hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11409856
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Hydrazinobenzoic acid hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It may be used for the preparation of 4-(3-(2-(tert-butoxycarbonylamino)ethyl)-5-hydroxy-1H-pyrazol-1-yl)benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydrazinobenzoic acid hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It may be used for the preparation of 4-(3-(2-(tert-butoxycarbonylamino)ethyl)-5-hydroxy-1H-pyrazol-1-yl)benzoic acid.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24589-77-3
Molecular Formula C7H9ClN2O2
Molecular Weight (g/mol) 188.61
MDL Number MFCD00039073
InChI Key XHLQMKQBCHYRLC-UHFFFAOYSA-N
Synonym 4-hydrazinobenzoic acid hydrochloride, 4-hydrazinylbenzoic acid hydrochloride, p-hydrazinobenzoic acid hydrochloride, 4-carboxyphenylhydrazine hydrochloride, benzoic acid, 4-hydrazino-, monohydrochloride, 4-carboxyphenylhydrazinium chloride, p-carboxyphenylhydrazine hydrochloride, ccris 7999, p-hydrazinobenzoic acid.hcl, 4-hydrazinobenzoic acid monohydrochloride
PubChem CID 90547
IUPAC Name 4-hydrazinylbenzoic acid;hydrochloride
SMILES [H+].[Cl-].NNC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 252°C to 254°C (decomposition)
Quantity 250 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 188.61
Percent Purity 98%
Chemical Name or Material 4-Hydrazinobenzoic acid hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'