missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-hydroxy-3-iodo-5-methoxybenzaldehyde, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Specifications
Assay Percent Range | 97% |
Linear Formula | IC6H2(OCH3)(OH)CHO |
Quantity | 25 g |
Formula Weight | 278.04 |
Percent Purity | 97% |
Chemical Name or Material | 5-Iodovanillin |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'