missing translation for 'onlineSavingsMsg'
Learn More
4-Hydroxy-3-methoxybenzonitrile, 98%, Thermo Scientific Chemicals Product Code.: 11409206

4-Hydroxy-3-methoxybenzonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11409206
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11409206

Brand: Thermo Scientific Alfa Aesar A18056.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4421-08-3
MDL Number MFCD00001820

Specifications

Melting Point 85°C to 89°C
Molecular Formula C8H5NO2
Quantity 100 g
UN Number UN3439
Synonym piperonylonitrile, benzo d 1,3 dioxole-5-carbonitrile, 3,4-methylenedioxy benzonitrile, 3,4-methylenedioxybenzonitrile, 2h-1,3-benzodioxole-5-carbonitrile, benzo-1,3-dioxole-5-carbonitrile, benzonitrile, 3,4-methylenedioxy, 2h-benzo d 1,3-dioxolane-5-carbonitrile, pubchem15358
InChI Key PKRWWZCDLJSJIF-UHFFFAOYSA-N
SMILES C1OC2=C(O1)C=C(C=C2)C#N
IUPAC Name 1,3-benzodioxole-5-carbonitrile
Molecular Weight (g/mol) 147.13
PubChem CID 78136
Formula Weight 149.15
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3-methoxybenzonitrile
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberG24N20 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18056.22,A18056.14
Lot Number10233459 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18056.22,A18056.14
Lot Number10159724 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18056.22,A18056.14
Lot Number10159473 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18056.22,A18056.14
Lot Number10138966 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18056.22,A18056.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Hydroxy-3-methoxybenzonitrile, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.