missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11463204
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Standard in chymotrypsin assays

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-82-0
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.12
MDL Number MFCD00007119
InChI Key QRYSWXFQLFLJTC-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitrobenzoic acid, 3-nitro-4-hydroxybenzoic acid, benzoic acid, 4-hydroxy-3-nitro, 3-nitro-4-hydrobenzoic acid, 4-hydroxy-3-nitro-benzoic acid, nhb, pubchem18981, acmc-1auqv, 3,4-nitrohydroxybenzoic acid, ksc489q0p
PubChem CID 12033
IUPAC Name 4-hydroxy-3-nitrobenzoic acid
SMILES OC(=O)C1=CC=C(O)C(=C1)[N+]([O-])=O

Specifications

Melting Point 182°C to 184°C
Quantity 25 g
Beilstein 2213134
Formula Weight 183.12
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3-nitrobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'