missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitrobenzonitrile, 98%, Thermo Scientific Chemicals

Product Code. 15411918
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3272-08-0
Molecular Formula C7H3N2O3
Molecular Weight (g/mol) 163.11
MDL Number MFCD00070775
InChI Key INBLGVOPOSGVTA-UHFFFAOYSA-M
Synonym 4-hydroxy-3-nitrobenzonitrile, 2-nitro-4-cyanophenol, benzonitrile, 4-hydroxy-3-nitro, 3-nitro-4-hydroxybenzonitrile, 4-cyano-2-nitrophenol, 4-hydroxy-3-nitro-benzonitrile, 4-hydroxy-3-nitrobenzenecarbonitrile, pubchem19690, acmc-1cq5c, ksc494q7b
PubChem CID 76758
IUPAC Name 4-cyano-2-nitrobenzen-1-olate
SMILES [O-]C1=CC=C(C=C1[N+]([O-])=O)C#N

Specifications

Melting Point 146°C to 148°C
Quantity 25 g
Beilstein 2048724
Formula Weight 164.12
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3-nitrobenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'