missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-4'-fluorobiphenyl, 98%, Thermo Scientific™

Product Code. 15478458
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 324-94-7
Molecular Formula C12H9FO
Molecular Weight (g/mol) 188.201
MDL Number MFCD01830385
InChI Key QSJNKJGPJVOGPK-UHFFFAOYSA-N
Synonym 4'-fluoro-1,1'-biphenyl-4-ol, 4-fluoro-4'-hydroxybiphenyl, 4-hydroxy-4'-fluorobiphenyl, 4-4-fluorophenyl phenol, 4'-fluoro-biphenyl-4-ol, 4'-fluoro 1,1'-biphenyl-4-ol, 4-4'-fluorophenyl phenol, 4'-fluorobiphenyl-1-ol, 1,1'-biphenyl-4-ol, 4'-fluoro, pubchem9057
PubChem CID 345420
IUPAC Name 4-(4-fluorophenyl)phenol
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)F)O

Specifications

Quantity 5 g
Formula Weight 188.2
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Hydroxy-4'-fluorobiphenyl
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'