missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-4'-iodobiphenyl, 98+%, Thermo Scientific Chemicals

Product Code. 11479627
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29558-78-9
Molecular Formula C12H9IO
Molecular Weight (g/mol) 296.107
MDL Number MFCD02258843
InChI Key WJXIAMCDWSUSEI-UHFFFAOYSA-N
Synonym 4-hydroxy-4'-iodobiphenyl, 4'-iodo-1,1'-biphenyl-4-ol, 4-4-iodophenyl phenol, 4-4'-iodophenyl phenol, 4'-iodobiphenyl-4-ol, acmc-20amgy, pubchem9061, 4-4?iodophenyl phenol, 4-4'-iodophenyl-phenol
PubChem CID 2064030
IUPAC Name 4-(4-iodophenyl)phenol
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)I)O

Specifications

Melting Point 196°C to 202°C
Quantity 25 g
Sensitivity Light sensitive
Formula Weight 296.11
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-4'-iodobiphenyl

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'