missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-4-methyl-2-pentanone, 99%, Thermo Scientific Chemicals

£27.34 - £224.12

Chemical Identifiers

CAS 123-42-2
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00004471
InChI Key SWXVUIWOUIDPGS-UHFFFAOYSA-N
Synonym diacetone alcohol, 4-hydroxy-4-methyl-2-pentanone, diacetonalkohol, diketone alcohol, diacetonalcohol, tyranton, acetonyldimethylcarbinol, diacetone-alcool, pyranton a, 2-methyl-2-pentanol-4-one
PubChem CID 31256
ChEBI CHEBI:55381
IUPAC Name 4-hydroxy-4-methylpentan-2-one
SMILES CC(=O)CC(C)(C)O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10626892
View Documents
Thermo Scientific Acros
121482500
250 mL Glass bottle
£27.34
250mL
Please to purchase this item. Need a web account? Register with us today!
10132490
View Documents Promotion Details
Thermo Scientific Acros
121480010
1 L Glass bottle
£66.43
1 L
Please to purchase this item. Need a web account? Register with us today!
10225450
View Documents
Thermo Scientific Acros
121480025
2.5 L Glass bottle
£76.68
2.50 L
Please to purchase this item. Need a web account? Register with us today!
11944861
View Documents
Thermo Scientific Acros
121480100
10 L Plastic drum
£224.12
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123-42-2
116.16
SWXVUIWOUIDPGS-UHFFFAOYSA-N
31256
4-hydroxy-4-methylpentan-2-one
C6H12O2
MFCD00004471
diacetone alcohol, 4-hydroxy-4-methyl-2-pentanone, diacetonalkohol, diketone alcohol, diacetonalcohol, tyranton, acetonyldimethylcarbinol, diacetone-alcool, pyranton a, 2-methyl-2-pentanol-4-one
CHEBI:55381
CC(=O)CC(C)(C)O

Specifications

123-42-2
Colorless
166.0°C
98.5% min. (GC)
(CH3)2C(OH)CH2COCH3
MFCD00004471
0.9385
Solubility in water: miscible. Other solubilities: miscible with alcohol,ether
CC(=O)CC(C)(C)O
116.16
CHEBI:55381
116.16
Liquid
-42.8°C
0.9385g/mL
58°C
C6H12O2
1.4223 to 1.4243
15,2967
diacetone alcohol, 4-hydroxy-4-methyl-2-pentanone, diacetonalkohol, diketone alcohol, diacetonalcohol, tyranton, acetonyldimethylcarbinol, diacetone-alcool, pyranton a, 2-methyl-2-pentanol-4-one
SWXVUIWOUIDPGS-UHFFFAOYSA-N
4-hydroxy-4-methylpentan-2-one
31256
3.22 mPa.s (20°C)
99%
4-Hydroxy-4-methyl-2-pentanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear eye protection/face protection.
If eye

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 204-626-7

Product Identifier
  • 4-Hydroxy-4-methyl-2-pentanone

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
  • Reproductive toxicity Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
  • H361-Suspected of damaging fertility or the unborn child.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Certificates

RUO – Research Use Only