missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-TEMPO, ≥98% (HPLC), Honeywell™ Fluka™

Product Code. 15645500
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 2226-96-2
Melting Point 69°C to 71°C
Color Orange-Red
Packaging Glass bottle
MDL Number MFCD00006478
Quantity 1 g
UN Number NONH for all modes of transport
Beilstein 1422990
Formula Weight 172.24g/mol
Percent Purity ≥98% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-TEMPO
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'