Learn More
4'-Hydroxyacetophenone, 98%, Thermo Scientific Chemicals
£29.65 - £116.58
Chemical Identifiers
CAS | 99-93-4 |
---|---|
Molecular Formula | C8H8O2 |
Molecular Weight (g/mol) | 136.15 |
MDL Number | MFCD00002359 |
InChI Key | TXFPEBPIARQUIG-UHFFFAOYSA-N |
Synonym | 4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone |
PubChem CID | 7469 |
ChEBI | CHEBI:28032 |
IUPAC Name | 1-(4-hydroxyphenyl)ethanone |
SMILES | CC(=O)C1=CC=C(O)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10785211
|
Thermo Scientific Acros
150400050 |
5 g | Glass bottle |
£29.65
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10448490
|
Thermo Scientific Acros
150401000 |
100 g | Plastic bottle |
£42.21
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10736371
|
Thermo Scientific Acros
150405000 |
500 g | Plastic bottle |
£116.58
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
99-93-4 | |
136.15 | |
TXFPEBPIARQUIG-UHFFFAOYSA-N | |
7469 | |
1-(4-hydroxyphenyl)ethanone |
C8H8O2 | |
MFCD00002359 | |
4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone | |
CHEBI:28032 | |
CC(=O)C1=CC=C(O)C=C1 |
Specifications
99-93-4 | |
100.0 | |
Beige to White | |
166°C | |
98% | |
HOC6H4COCH3 | |
08, 87 | |
Solubility in water: 10g/L (22°C) | |
CC(=O)C1=CC=C(O)C=C1 | |
136.15 | |
CHEBI:28032 | |
98% | |
4'-Hydroxyacetophenone, 98% |
97.5 | |
107°C to 111°C | |
147°C to 148°C (3.0 mmHg) | |
Authentic | |
C8H8O2 | |
MFCD00002359 | |
4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone | |
TXFPEBPIARQUIG-UHFFFAOYSA-N | |
1-(4-hydroxyphenyl)ethanone | |
7469 | |
136.15 | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-802-8
missing translation for 'rtecsNumber' : AM8750000
missing translation for 'tsca' : TSCA
- 4'-Hydroxyacetophenone
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only