missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
4-Hydroxybenzaldehyde is used as a chemical that maintains bactericidal activity when tested against certain bacteria strains.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hydroxybenzaldehyde is used as a chemical that maintains bactericidal activity when tested against certain bacteria strains.
Solubility
Soluble in water(13.8g/L), ether, alcohol, and acetone.
Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
4-Hydroxybenzaldehyde is used as a chemical that maintains bactericidal activity when tested against certain bacteria strains.
Solubility
Soluble in water(13.8g/L), ether, alcohol, and acetone.
Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 115°C to 119°C |
Density | 1.129 |
Boiling Point | 191°C (50 mmHg) |
Flash Point | 174°C (345°F) |
Quantity | 25 g |
Beilstein | 471352 |
Sensitivity | Air Sensitive |
Merck Index | 14,4811 |
Solubility Information | Soluble in water(13.8g/L),ether,alcohol,and acetone. |
Formula Weight | 122.12 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'