Learn More
4-Hydroxybenzoic acid, 99%, Thermo Scientific Chemicals
£31.16 - £155.78
Chemical Identifiers
CAS | 99-96-7 |
---|---|
Molecular Formula | C7H6O3 |
Molecular Weight (g/mol) | 138.12 |
MDL Number | MFCD00002547 |
InChI Key | FJKROLUGYXJWQN-UHFFFAOYSA-N |
Synonym | p-hydroxybenzoic acid, 4-carboxyphenol, benzoic acid, 4-hydroxy, p-salicylic acid, benzoic acid, p-hydroxy, para-hydroxybenzoic acid, 4-hydroxybenzoesaeure, p-carboxyphenol, 4-hydroxy benzoic acid, p-oxybenzoesaure german |
PubChem CID | 135 |
ChEBI | CHEBI:30763 |
IUPAC Name | 4-hydroxybenzoic acid |
SMILES | OC(=O)C1=CC=C(O)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11424694
|
Thermo Scientific Alfa Aesar
A13700.30 |
250 g |
£31.16
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414694
|
Thermo Scientific Alfa Aesar
A13700.0B |
1000 g |
£39.60
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434694
|
Thermo Scientific Alfa Aesar
A13700.0I |
5000 g |
£155.78
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Hydroxybenzoic acid is used as an intermediate for dyes, antiseptics and active pharmaceutical ingredients. It is also used as a food preservative, corrosion inhibitor, anti-oxidant and an emulsifier. It reacts with 6-hydroxynaphthalene-2-carboxylic acid to prepare aromatic polyester called Vectran fiber. It acts as a precursor to prepare p-hydroxyphenylbenzoate, p-acetoxybenzoyl chloride and 4,4'-dihydroxybenzophenone. Further, it is used in paint additives, coating additives, processing aids, electrical and electronic products. In addition to this, it acts as a preservative in cosmetics and some ophthalmic solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Hydroxybenzoic acid is used as an intermediate for dyes, antiseptics and active pharmaceutical ingredients. It is also used as a food preservative, corrosion inhibitor, anti-oxidant and an emulsifier. It reacts with 6-hydroxynaphthalene-2-carboxylic acid to prepare aromatic polyester called Vectran fiber. It acts as a precursor to prepare p-hydroxyphenylbenzoate, p-acetoxybenzoyl chloride and 4,4′-dihydroxybenzophenone. Further, it is used in paint additives, coating additives, processing aids, electrical and electronic products. In addition to this, it acts as a preservative in cosmetics and some ophthalmic solutions.
Solubility
Soluble in water, alcohol, ethanol, ether, n-butanol and acetone. Slightly soluble in chloroform. Insoluble in carbon disulfide.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
99-96-7 | |
138.12 | |
FJKROLUGYXJWQN-UHFFFAOYSA-N | |
135 | |
4-hydroxybenzoic acid |
C7H6O3 | |
MFCD00002547 | |
p-hydroxybenzoic acid, 4-carboxyphenol, benzoic acid, 4-hydroxy, p-salicylic acid, benzoic acid, p-hydroxy, para-hydroxybenzoic acid, 4-hydroxybenzoesaeure, p-carboxyphenol, 4-hydroxy benzoic acid, p-oxybenzoesaure german | |
CHEBI:30763 | |
OC(=O)C1=CC=C(O)C=C1 |
Specifications
99-96-7 | |
199°C (390°F) | |
C7H6O3 | |
970950 | |
p-hydroxybenzoic acid, 4-carboxyphenol, benzoic acid, 4-hydroxy, p-salicylic acid, benzoic acid, p-hydroxy, para-hydroxybenzoic acid, 4-hydroxybenzoesaeure, p-carboxyphenol, 4-hydroxy benzoic acid, p-oxybenzoesaure german | |
FJKROLUGYXJWQN-UHFFFAOYSA-N | |
4-hydroxybenzoic acid | |
135 | |
138.12 | |
4-Hydroxybenzoic Acid |
213°C to 218°C | |
99% | |
MFCD00002547 | |
14,4812 | |
Soluble in water,alcohol,ethanol,ether,n-butanol and acetone. Slightly soluble in chloroform. Insoluble in carbon disulfide. | |
OC(=O)C1=CC=C(O)C=C1 | |
138.12 | |
CHEBI:30763 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335
P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
H318-H335
missing translation for 'einecsNumber' : 202-804-9
missing translation for 'rtecsNumber' : DH1925000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 4-Hydroxybenzoic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : p-Hydroxybenzoic acid
RUO – Research Use Only