missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzophenone, 98+%, Thermo Scientific Chemicals

Product Code. 10285250
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1137-42-4
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00002355
InChI Key NPFYZDNDJHZQKY-UHFFFAOYSA-N
Synonym 4-hydroxybenzophenone, 4-benzoylphenol, 4-hydroxyphenyl phenyl methanone, p-benzoylphenol, p-hydroxybenzophenone, methanone, 4-hydroxyphenyl phenyl, 4-hydroxyphenyl phenylmethanone, benzophenone, 4-hydroxy, unii-04r2lws0ms, 4'-hydroxybenzophenone
PubChem CID 14347
ChEBI CHEBI:34421
IUPAC Name (4-hydroxyphenyl)-phenylmethanone
SMILES OC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 132.0°C to 135.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula HOC6H4COC6H5
Quantity 25 g
Beilstein 08,II,184
Solubility Information Solubility in water: insoluble
Formula Weight 198.22
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 4-Hydroxybenzophenone, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'