missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymercury)benzoic Acid Sodium Salt, 98%, Thermo Scientific Chemicals

Product Code. 10576592
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-85-2
Molecular Formula C7H5HgNaO3
Molecular Weight (g/mol) 360.69
MDL Number MFCD00002527
InChI Key BERBQUNVJGVZCX-UHFFFAOYSA-M
Synonym sodium 4-carboxylatophenyl mercury hydrate, sodium hydrate p-mercuribenzoate
PubChem CID 23667696
IUPAC Name sodium;(4-carboxylatophenyl)mercury;hydrate
SMILES C1=CC(=CC=C1C(=O)[O-])[Hg].O.[Na+]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Hg)
Packaging Glass bottle
Quantity 1 g
Beilstein 16, 969
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 360.69
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Hydroxymercury)benzoic acid sodium salt
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-(Hydroxymercury)benzoic acid sodium salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'