Learn More
4-Hydroxyphenethyl alcohol, 98%, Thermo Scientific Chemicals
£40.30 - £156.00
Chemical Identifiers
CAS | 501-94-0 |
---|---|
Molecular Formula | C8H10O2 |
Molecular Weight (g/mol) | 138.17 |
MDL Number | MFCD00002902 |
InChI Key | YCCILVSKPBXVIP-UHFFFAOYSA-N |
Synonym | tyrosol, 2-4-hydroxyphenyl ethanol, 4-hydroxyphenethyl alcohol, 4-2-hydroxyethyl phenol, 4-hydroxyphenylethanol, 4-hydroxybenzeneethanol, p-hydroxyphenethyl alcohol, benzeneethanol, 4-hydroxy, p-tyrosol, p-hydroxyphenylethanol |
PubChem CID | 10393 |
ChEBI | CHEBI:1879 |
IUPAC Name | 4-(2-hydroxyethyl)phenol |
SMILES | C1=CC(=CC=C1CCO)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10114060
|
Thermo Scientific Acros
182500050 |
5 g | Glass bottle |
£40.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10686192
|
Thermo Scientific Acros
182500250 |
25 g | Glass bottle |
£156.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
501-94-0 | |
138.17 | |
YCCILVSKPBXVIP-UHFFFAOYSA-N | |
10393 | |
4-(2-hydroxyethyl)phenol |
C8H10O2 | |
MFCD00002902 | |
tyrosol, 2-4-hydroxyphenyl ethanol, 4-hydroxyphenethyl alcohol, 4-2-hydroxyethyl phenol, 4-hydroxyphenylethanol, 4-hydroxybenzeneethanol, p-hydroxyphenethyl alcohol, benzeneethanol, 4-hydroxy, p-tyrosol, p-hydroxyphenylethanol | |
CHEBI:1879 | |
C1=CC(=CC=C1CCO)O |
Specifications
501-94-0 | |
100.0 | |
White to Beige | |
97.5% min. (GC) | |
HOC6H4CH2CH2OH | |
06, IV, 5936 | |
tyrosol, 2-4-hydroxyphenyl ethanol, 4-hydroxyphenethyl alcohol, 4-2-hydroxyethyl phenol, 4-hydroxyphenylethanol, 4-hydroxybenzeneethanol, p-hydroxyphenethyl alcohol, benzeneethanol, 4-hydroxy, p-tyrosol, p-hydroxyphenylethanol | |
YCCILVSKPBXVIP-UHFFFAOYSA-N | |
4-(2-hydroxyethyl)phenol | |
10393 | |
138.17 | |
Crystalline Powder or Crystals |
97.5 | |
89.0°C to 92.0°C | |
Authentic | |
C8H10O2 | |
MFCD00002902 | |
15, 10022 | |
Solubility in water: 124g/L (25°C). Other solubilities: soluble in methanol,ethanol,acetone,chloroform,,diethyl ether,petroleum ether | |
C1=CC(=CC=C1CCO)O | |
138.17 | |
CHEBI:1879 | |
98% | |
4-Hydroxyphenethyl alcohol, 98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 207-930-8
missing translation for 'tsca' : TSCA
- 4-Hydroxyphenethyl alcohol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only