missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenyl benzoate, 98%, Thermo Scientific Chemicals

Product Code. 11397366
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2444-19-1
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00053304
InChI Key JFAXJRJMFOACBO-UHFFFAOYSA-N
Synonym hydroquinone monobenzoate, p-hydroxyphenyl benzoate, unii-zcy7o225cp, 4-benzoyloxyphenol, 1,4-diphenol, 1-benzoate, benzoic acid 4-hydroxyphenyl ester, 4-hydroxyphenyl benzoate, zcy7o225cp, 4-hydroxyphenylbenzoate, 4-hydroxyphenylbenzoat
PubChem CID 75549
IUPAC Name (4-hydroxyphenyl) benzoate
SMILES OC1=CC=C(OC(=O)C2=CC=CC=C2)C=C1

Specifications

Melting Point 164°C to 167°C
Quantity 5 g
Beilstein 2050596
Formula Weight 214.22
Percent Purity 98%
Chemical Name or Material 4-Hydroxyphenyl benzoate
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.