missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylacetamide, 99%, Thermo Scientific Chemicals

Product Code. 11402328
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17194-82-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00017145
InChI Key YBPAYPRLUDCSEY-UHFFFAOYSA-N
Synonym 4-hydroxyphenylacetamide, 2-4-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, 4-hydroxybenzeneacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxyphenyl acetamide, unii-0hy0n4itn4
PubChem CID 86986
IUPAC Name 2-(4-hydroxyphenyl)acetamide
SMILES C1=CC(=CC=C1CC(=O)N)O

Specifications

Melting Point 173°C to 175°C
Quantity 250 g
Beilstein 2085984
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 4-Hydroxyphenylacetamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'