Learn More
4-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals
£30.80 - £370.00
Chemical Identifiers
CAS | 156-38-7 |
---|---|
Molecular Formula | C8H8O3 |
Molecular Weight (g/mol) | 152.149 |
MDL Number | MFCD00004347 |
InChI Key | XQXPVVBIMDBYFF-UHFFFAOYSA-N |
Synonym | 4-hydroxyphenylacetic acid, 2-4-hydroxyphenyl acetic acid, p-hydroxyphenylacetic acid, 4-hydroxyphenyl acetic acid, 4-hydroxyphenylacetate, 4-hydroxybenzeneacetic acid, benzeneacetic acid, 4-hydroxy, p-hydroxyphenyl acetic acid, parahydroxy phenylacetic acid, 4-carboxymethylphenol |
PubChem CID | 127 |
ChEBI | CHEBI:18101 |
IUPAC Name | 2-(4-hydroxyphenyl)acetic acid |
SMILES | C1=CC(=CC=C1CC(=O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11497257
|
Thermo Scientific Alfa Aesar
A15018.14 |
25 g |
£30.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11407267
|
Thermo Scientific Alfa Aesar
A15018.22 |
100 g |
£81.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11417267
|
Thermo Scientific Alfa Aesar
A15018.36 |
500 g |
£370.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a reagent in the acylation of phenols and amines. It is also used for fluorometric determination of oxidative enzymes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a reagent in the acylation of phenols and amines. It is also used for fluorometric determination of oxidative enzymes.
Solubility
Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Chemical Identifiers
156-38-7 | |
152.149 | |
XQXPVVBIMDBYFF-UHFFFAOYSA-N | |
127 | |
2-(4-hydroxyphenyl)acetic acid |
C8H8O3 | |
MFCD00004347 | |
4-hydroxyphenylacetic acid, 2-4-hydroxyphenyl acetic acid, p-hydroxyphenylacetic acid, 4-hydroxyphenyl acetic acid, 4-hydroxyphenylacetate, 4-hydroxybenzeneacetic acid, benzeneacetic acid, 4-hydroxy, p-hydroxyphenyl acetic acid, parahydroxy phenylacetic acid, 4-carboxymethylphenol | |
CHEBI:18101 | |
C1=CC(=CC=C1CC(=O)O)O |
Specifications
156-38-7 | |
C8H8O3 | |
1448766 | |
Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water. | |
C1=CC(=CC=C1CC(=O)O)O | |
152.149 | |
CHEBI:18101 | |
99% |
148°C to 150°C | |
MFCD00004347 | |
4-hydroxyphenylacetic acid, 2-4-hydroxyphenyl acetic acid, p-hydroxyphenylacetic acid, 4-hydroxyphenyl acetic acid, 4-hydroxyphenylacetate, 4-hydroxybenzeneacetic acid, benzeneacetic acid, 4-hydroxy, p-hydroxyphenyl acetic acid, parahydroxy phenylacetic acid, 4-carboxymethylphenol | |
XQXPVVBIMDBYFF-UHFFFAOYSA-N | |
2-(4-hydroxyphenyl)acetic acid | |
127 | |
152.15 | |
4-Hydroxyphenylacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 205-851-3
missing translation for 'rtecsNumber' : AI2680000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only