missing translation for 'onlineSavingsMsg'
Learn More

4-iodo-2-methyl-1H-imidazole, Thermo Scientific™

Product Code. 10513232
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 73746-45-9
Molecular Formula C4H5IN2
Molecular Weight (g/mol) 208.00
MDL Number MFCD02684335
InChI Key SEDSLMWYUQACGR-UHFFFAOYSA-N
PubChem CID 2794623
IUPAC Name 5-iodo-2-methyl-1H-imidazole
SMILES CC1=NC=C(I)N1

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Synonym 4-iodo-2-methyl-1h-imidazole, 2-methyl-4 5-iodo-1 h-imidazole, 4-iodo-2-methylimidazole, 2-methyl-4-iodo-1h-imidazole, 4-iodo-2-methyl-3h-imidazole, 1h-imidazole, 4-iodo-2-methyl, 2-methyl-4-jodimidazol, ksc638k7d, 1h-imidazole,4-iodo-2-methyl, 1h-imidazole,5-iodo-2-methyl
Formula Weight 208.0016
Chemical Name or Material 4-iodo-2-methyl-1H-imidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'