missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodo-3,5-bis-(trifluoromethyl)-1H-pyrazole, 97%, Thermo Scientific™
£232.65 - £1126.03
Chemical Identifiers
CAS | 1239511-06-8 |
---|---|
Molecular Formula | C5HF6IN2 |
Molecular Weight (g/mol) | 329.97 |
MDL Number | MFCD11867872 |
InChI Key | OTYORVJCWWESOF-UHFFFAOYSA-N |
Synonym | 4-iodo-3,5-bis trifluoromethyl-1h-pyrazole, 3,5-bis trifluoromethyl-4-iodo-1h-pyrazole |
PubChem CID | 45933690 |
IUPAC Name | 4-iodo-3,5-bis(trifluoromethyl)-1H-pyrazole |
SMILES | FC(F)(F)C1=C(I)C(=NN1)C(F)(F)F |
Description
Chemical Identifiers
1239511-06-8 | |
329.97 | |
OTYORVJCWWESOF-UHFFFAOYSA-N | |
45933690 | |
FC(F)(F)C1=C(I)C(=NN1)C(F)(F)F |
C5HF6IN2 | |
MFCD11867872 | |
4-iodo-3,5-bis trifluoromethyl-1h-pyrazole, 3,5-bis trifluoromethyl-4-iodo-1h-pyrazole | |
4-iodo-3,5-bis(trifluoromethyl)-1H-pyrazole |
Specifications
1239511-06-8 | |
96% min. (GC) | |
C5HF6IN2 | |
4-iodo-3,5-bis trifluoromethyl-1h-pyrazole, 3,5-bis trifluoromethyl-4-iodo-1h-pyrazole | |
FC(F)(F)C1=C(I)C(=NN1)C(F)(F)F | |
329.97 | |
329.97 | |
4-Iodo-3, 5-bis-(trifluoromethyl)-1H-pyrazole |
125°C to 126°C | |
Glass Bottle | |
MFCD11867872 | |
OTYORVJCWWESOF-UHFFFAOYSA-N | |
4-iodo-3,5-bis(trifluoromethyl)-1H-pyrazole | |
45933690 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4-Iodo-3,5-bis-(trifluoromethyl)-1H-pyrazole
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only