missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodo-L-phenylalanine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
4-Iodo-L-phenylalanine may be used in protein engineering as a model unnatural α amino acid to alter primary amino acid composition via the opal (UGA) codon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Iodo-L-phenylalanine may be used in protein engineering as a model unnatural α amino acid to alter primary amino acid composition via the opal (UGA) codon.
Solubility
Partly miscible with water.
Notes
Light sensitive. Incompatible with oxidizing agents.
4-Iodo-L-phenylalanine may be used in protein engineering as a model unnatural α amino acid to alter primary amino acid composition via the opal (UGA) codon.
Solubility
Partly miscible with water.
Notes
Light sensitive. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Beilstein | 4411317 |
Solubility Information | Partly miscible with water. |
Formula Weight | 291.09 |
Percent Purity | 95% |
Quantity | 250 mg |
Optical Rotation | −6.5° (c=1 in Acetic Acid:Water (4:1)) |
Sensitivity | Light sensitive |
Chemical Name or Material | 4-Iodo-L-phenylalanine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'