missing translation for 'onlineSavingsMsg'
Learn More

4'-Iodoacetophenone, 98%

Product Code. 11492818
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11492818

Brand: Thermo Scientific Alfa Aesar A16794.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13329-40-3 | C8H7IO | 246.047 g/mol

Synthesis of quinoline-based potential anticancer agents. Substrate for palladium-catalyzed coupling reactions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis of quinoline-based potential anticancer agents. Substrate for palladium-catalyzed coupling reactions

Solubility
Easily soluble in water.

Notes
Light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13329-40-3
Molecular Formula C8H7IO
Molecular Weight (g/mol) 246.047
MDL Number MFCD00045320
InChI Key JZJWCDQGIPQBAO-UHFFFAOYSA-N
Synonym 4'-iodoacetophenone, 1-4-iodophenyl ethanone, 4-iodoacetophenone, 1-4-iodophenyl ethan-1-one, p-iodoacetophenone, ethanone, 1-4-iodophenyl, 1-4-iodo-phenyl-ethanone, 1-acetyl-4-iodobenzene, 4-iodo acetophenone, 4;-iodoacetophenone
PubChem CID 72869
IUPAC Name 1-(4-iodophenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)I

Specifications

Melting Point 82°C to 84°C
Quantity 5 g
Beilstein 1857412
Sensitivity Light sensitive
Solubility Information Easily soluble in water.
Formula Weight 246.05
Percent Purity 98%
Chemical Name or Material 4'-Iodoacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.