missing translation for 'onlineSavingsMsg'
Learn More
4-Iodoaniline, 98%, Thermo Scientific Chemicals Product Code.: 11485027

4-Iodoaniline, 98%, Thermo Scientific Chemicals

Product Code. 11485027
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11485027

Brand: Thermo Scientific Alfa Aesar B21936.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-dehydroanilinium ion, a stable distonic isomer of ionized aniline, by collision-activated dissociation of protonated 4-iodoaniline. Palladium-catalyzed carbonylation of 4-iodoaniline gave the corresponding four- armed star polybenzamides. 4-Iodoaniline is a iodine substituted aniline that is widely used as chemical intermediates in the manufacturing of dyes and drugs. 4-Iodoaniline was used to prepare phenyl functionalized graphene oxide (I-Ph-GO).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-dehydroanilinium ion, a stable distonic isomer of ionized aniline, by collision-activated dissociation of protonated 4-iodoaniline. Palladium-catalyzed carbonylation of 4-iodoaniline gave the corresponding four- armed star polybenzamides. 4-Iodoaniline is a iodine substituted aniline that is widely used as chemical intermediates in the manufacturing of dyes and drugs. 4-Iodoaniline was used to prepare phenyl functionalized graphene oxide (I-Ph-GO).

Solubility
Slightly soluble in water.Soluble in chloroform, methanol.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-37-4
Molecular Formula C6H6IN
Molecular Weight (g/mol) 219.025
MDL Number MFCD00007848
InChI Key VLVCDUSVTXIWGW-UHFFFAOYSA-N
Synonym p-iodoaniline, benzenamine, 4-iodo, 4-iodobenzenamine, aniline, p-iodo, p-aminophenyl iodide, aniline, 4-iodo, 4-iodo-aniline, 4-iodophenylamine, 4-iodo-benzenamine, 4-iodo-phenylamine
PubChem CID 10893
IUPAC Name 4-iodoaniline
SMILES C1=CC(=CC=C1N)I

Specifications

Melting Point 61°C to 64°C
Quantity 50 g
Beilstein 774101
Sensitivity Light sensitive
Merck Index 14,5027
Solubility Information Slightly soluble in water.Soluble in chloroform,methanol.
Formula Weight 219.03
Percent Purity 98%
Chemical Name or Material 4-Iodoaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Iodoaniline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.