missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobutyronitrile, 97%

Product Code. 10106303
1 g, Glass bottle
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 6727-73-7 | C4H6IN | 195 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6727-73-7
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.8260g/mL
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C4H6IN
Linear Formula I(CH2)3CN
Refractive Index 1.5350 to 1.5370
MDL Number MFCD00039501
Quantity 1 g
Specific Gravity 1.826
Synonym butanenitrile, 4-iodo, g-iodobutyronitrile, 4-iodobutyronitrile
InChI Key CWINCVBWHUGBEB-UHFFFAOYSA-N
SMILES C(CC#N)CI
IUPAC Name 4-iodobutanenitrile
Molecular Weight (g/mol) 195
PubChem CID 285648
Formula Weight 195
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Iodobutyronitrile
Show More Show Less
compliance-icons
Product Identifier
  • 4-Iodobutyronitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Iodobutyronitrile, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.