missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenetole, 97%, Thermo Scientific Chemicals

Product Code. 11430207
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 699-08-1
Molecular Formula C8H9IO
Molecular Weight (g/mol) 248.063
MDL Number MFCD00061124
InChI Key VSIIHWOJPSSIDI-UHFFFAOYSA-N
Synonym 4-iodophenetole, p-iodophenetole, 1-ethoxy-4-iodo-benzene, 4-ethoxyiodobenzene, 4-ethoxy-4-iodobenzene, benzene, 1-ethoxy-4-iodo, 4-iodophenyl ethyl ether, phenetole, p-iodo, p-ethoxyiodobenzene, ethyl 4-iodophenyl ether
PubChem CID 220700
IUPAC Name 1-ethoxy-4-iodobenzene
SMILES CCOC1=CC=C(C=C1)I

Specifications

Melting Point 26°C to 29°C
Boiling Point 133°C to 134°C (19 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.597
Quantity 100 g
Beilstein 2042067
Sensitivity Light sensitive
Formula Weight 248.06
Percent Purity 97%
Chemical Name or Material 4-Iodophenetole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'