missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodophenylacetic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4-Iodophenylacetic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in suzuki coupling reaction transport.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Iodophenylacetic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in suzuki coupling reaction transport.
Solubility
Insoluble in water.
Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
4-Iodophenylacetic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in suzuki coupling reaction transport.
Solubility
Insoluble in water.
Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 136°C to 140°C |
Quantity | 5 g |
Beilstein | 1940558 |
Sensitivity | Light sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 262.05 |
Percent Purity | 97% |
Chemical Name or Material | 4-Iodophenylacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'