Learn More
4-Isocyanato-4-thien-2-yltetrahydropyran, 95%, Thermo Scientific™
£31.00 - £90.00
Chemical Identifiers
CAS | 926921-63-3 |
---|---|
Molecular Formula | C10H11NO2S |
Molecular Weight (g/mol) | 209.26 |
MDL Number | MFCD09879931 |
InChI Key | CDJWJYYYKMFRFC-UHFFFAOYSA-N |
Synonym | 4-isocyanato-4-thiophen-2-yl oxane, 4-isocyanato-4-thien-2-yl tetrahydro-2h-pyran, 4-thien-2-yltetrahydro-2h-pyran-4-yl isocyanate, 4-isocyanato-4-thien-2-yltetrahydropyran, 2-4-isocyanatotetrahydro-2h-pyran-4-yl thiophene, 4-thien-2-yl tetrahydro-2h-pyran-4-yl isocyanate, 4-isocyanato-4-thiophen-2-yltetrahydropyran, 4-2-thienyl-2h-3,4,5,6-tetrahydropyran-4-isocyanate |
PubChem CID | 24229667 |
IUPAC Name | 4-isocyanato-4-thiophen-2-yloxane |
SMILES | O=C=NC1(CCOCC1)C1=CC=CS1 |
Description
Chemical Identifiers
926921-63-3 | |
209.26 | |
CDJWJYYYKMFRFC-UHFFFAOYSA-N | |
24229667 | |
O=C=NC1(CCOCC1)C1=CC=CS1 |
C10H11NO2S | |
MFCD09879931 | |
4-isocyanato-4-thiophen-2-yl oxane, 4-isocyanato-4-thien-2-yl tetrahydro-2h-pyran, 4-thien-2-yltetrahydro-2h-pyran-4-yl isocyanate, 4-isocyanato-4-thien-2-yltetrahydropyran, 2-4-isocyanatotetrahydro-2h-pyran-4-yl thiophene, 4-thien-2-yl tetrahydro-2h-pyran-4-yl isocyanate, 4-isocyanato-4-thiophen-2-yltetrahydropyran, 4-2-thienyl-2h-3,4,5,6-tetrahydropyran-4-isocyanate | |
4-isocyanato-4-thiophen-2-yloxane |
Specifications
926921-63-3 | |
100.0 | |
C10H11NO2S | |
4-isocyanato-4-thiophen-2-yl oxane, 4-isocyanato-4-thien-2-yl tetrahydro-2h-pyran, 4-thien-2-yltetrahydro-2h-pyran-4-yl isocyanate, 4-isocyanato-4-thien-2-yltetrahydropyran, 2-4-isocyanatotetrahydro-2h-pyran-4-yl thiophene, 4-thien-2-yl tetrahydro-2h-pyran-4-yl isocyanate, 4-isocyanato-4-thiophen-2-yltetrahydropyran, 4-2-thienyl-2h-3,4,5,6-tetrahydropyran-4-isocyanate | |
O=C=NC1(CCOCC1)C1=CC=CS1 | |
209.26 | |
209.27 | |
4-Isocyanato-4-thien-2-yltetrahydropyran |
94.5 | |
95% | |
MFCD09879931 | |
CDJWJYYYKMFRFC-UHFFFAOYSA-N | |
4-isocyanato-4-thiophen-2-yloxane | |
24229667 | |
95% |
Safety and Handling
GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
Your input is important to us. Please complete this form to provide feedback related to the content on this product.