missing translation for 'onlineSavingsMsg'
Learn More

4-Isocyanato-5-methyl-1-phenyl-1H-pyrazole 97+%, Thermo Scientific™

Product Code. 10770904
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 799283-97-9
Molecular Formula C11H9N3O
Molecular Weight (g/mol) 199.213
InChI Key IIFVDOYRTLEGHQ-UHFFFAOYSA-N
Synonym 4-isocyanato-5-methyl-1-phenyl-1h-pyrazole, 5-methyl-1-phenylpyrazol-4-isocyanate, 1h-pyrazole,4-isocyanato-5-methyl-1-phenyl, 5-methyl-1-phenyl-1h-pyrazol-4-yl isocyanate, 4-isocyanato-5-methyl-1h-pyrazol-4-yl benzene
PubChem CID 7164551
IUPAC Name 4-isocyanato-5-methyl-1-phenylpyrazole
SMILES CC1=C(C=NN1C2=CC=CC=C2)N=C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 199.21
Percent Purity 97%
Chemical Name or Material 4-Isocyanato-5-methyl-1-phenyl-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'