Learn More
4-Isopropyl-3-methylaniline hydrochloride, 97%, Thermo Scientific™
£56.40 - £95.00
Chemical Identifiers
CAS | 4534-11-6 |
---|---|
Molecular Formula | C10H16ClN |
Molecular Weight (g/mol) | 185.70 |
MDL Number | MFCD00156562 |
InChI Key | VPOAQDSAPBSOLW-UHFFFAOYSA-N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10304732
|
Thermo Scientific Maybridge
SB01749EA |
10 g |
£56.40
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10215314
|
Thermo Scientific Maybridge
SB01749EB |
25 g | |||||||
Description
Chemical Identifiers
4534-11-6 | |
185.70 | |
VPOAQDSAPBSOLW-UHFFFAOYSA-N |
C10H16ClN | |
MFCD00156562 |
Specifications
4534-11-6 | |
95% | |
C10H16ClN | |
3-methyl-4-isopropylaniline hydrochloride, 4-isopropyl-3-methylaniline hydrochloride, 4-isopropyl-m-toluidinium chloride, benzenamine, 3-methyl-4-1-methylethyl-, hydrochloride, acmc-20a42e, 3-methyl-4-propan-2-ylaniline hydrochloride, 4-isopropyl-3-methylbenzenamine hydrochloride, 3-methyl-4-methylethyl phenylamine, chloride, 3-methyl-4-propan-2-yl aniline hydrochloride, benzenamine, 3-?methyl-?4-? 1-?methylethyl ?-?, hy | |
CC1=C(C=CC(=C1)N)C(C)C.Cl | |
185.70 | |
185.7 | |
4-Isopropyl-3-methylaniline hydrochloride |
95.0 | |
Amber Glass Bottle | |
MFCD00156562 | |
VPOAQDSAPBSOLW-UHFFFAOYSA-N | |
3-methyl-4-propan-2-ylaniline;hydrochloride | |
2796270 | |
95% |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning