missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropyl-3-methylaniline hydrochloride, 97%, Thermo Scientific™

Product Code. 11408637
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Isopropyl-3-methylaniline hydrochloride is used to synthesis of fine chemical, organic intermediates.

Solubility
Slightly soluble in water (9.9 g/L)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4534-11-6
Molecular Formula C10H16ClN
Molecular Weight (g/mol) 185.70
MDL Number MFCD00156562
InChI Key VPOAQDSAPBSOLW-UHFFFAOYSA-N
Synonym 3-methyl-4-isopropylaniline hydrochloride, 4-isopropyl-3-methylaniline hydrochloride, 4-isopropyl-m-toluidinium chloride, benzenamine, 3-methyl-4-1-methylethyl-, hydrochloride, acmc-20a42e, 3-methyl-4-propan-2-ylaniline hydrochloride, 4-isopropyl-3-methylbenzenamine hydrochloride, 3-methyl-4-methylethyl phenylamine, chloride, 3-methyl-4-propan-2-yl aniline hydrochloride, benzenamine, 3-?methyl-?4-? 1-?methylethyl ?-?, hy
PubChem CID 2796270
IUPAC Name 3-methyl-4-propan-2-ylaniline;hydrochloride
SMILES CC1=C(C=CC(=C1)N)C(C)C.Cl

Specifications

Melting Point 222°C to 225°C
Quantity 5 g
Solubility Information Slightly soluble in water (9.9g/L)
Formula Weight 185.7
Percent Purity 97%
Chemical Name or Material 4-Isopropyl-3-methylaniline hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.