missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzenesulfonyl chloride, 96%, Thermo Scientific Chemicals

Product Code. 11388425
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54997-90-9
Molecular Formula C9H11ClO2S
Molecular Weight (g/mol) 218.70
MDL Number MFCD00041510
InChI Key CETRNHJIXGITKR-UHFFFAOYSA-N
Synonym 4-isopropylbenzenesulfonyl chloride, 4-isopropylbenzene-1-sulfonyl chloride, 4-isopropylbenzenesulfonylchloride, 4-propan-2-yl benzene-1-sulfonyl chloride, 4-isopropyl benzenesulfonyl chloride, 4-isopropyl-benzenesulfonyl chloride, 4-propan-2-yl benzenesulfonyl chloride, 4-isopropylbenzensulfonylchloride, 4-isopropylphenylsulfonyl chloride, p-isopropylbenzenesulfonyl chloride
PubChem CID 2734882
IUPAC Name 4-propan-2-ylbenzenesulfonyl chloride
SMILES CC(C)C1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Density 1.22
Boiling Point 142°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.539
Quantity 25 g
UN Number UN3265
Beilstein 1103862
Sensitivity Moisture sensitive
Formula Weight 218.7
Percent Purity 96%
Chemical Name or Material 4-Isopropylbenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'