missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenol, 98%, Thermo Scientific Chemicals

Product Code. 10092130
Click to view available options
Quantity:
5 g
100 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-89-8
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00002372
InChI Key YQUQWHNMBPIWGK-UHFFFAOYSA-N
Synonym 4-isopropylphenol, p-isopropylphenol, p-cumenol, australol, phenol, 4-1-methylethyl, 4-1-methylethyl phenol, 1-hydroxy-4-isopropylbenzene, 4-propan-2-yl phenol, phenol, p-isopropyl, para-isopropylphenol
PubChem CID 7465
IUPAC Name 4-propan-2-ylphenol
SMILES CC(C)C1=CC=C(C=C1)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 59.0°C to 62.0°C
Boiling Point 212.0°C to 212.4°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHC6H4OH
Quantity 5 g
Beilstein 06,505
Solubility Information Solubility in water: .. Other solubilities: soluble in 10% sodium hydroxide,soluble in alcohol and ethers
Formula Weight 136.19
Percent Purity 98%
Physical Form Crystalline Mass or Crystals
Chemical Name or Material 4-Isopropylphenol, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'