missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Isopropylphenol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
4-Isopropylphenol was used in the synthesis of vinyl compounds. It is used as a flavor & fragrance agent in cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Isopropylphenol was used in the synthesis of vinyl compounds. It is used as a flavor & fragrance agent in cosmetics.
Solubility
Slightly soluble in water, soluble in ethanol and ethyl ether.
Notes
Incompatible with oxidizing agents.
4-Isopropylphenol was used in the synthesis of vinyl compounds. It is used as a flavor & fragrance agent in cosmetics.
Solubility
Slightly soluble in water, soluble in ethanol and ethyl ether.
Notes
Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 59°C to 61°C |
Boiling Point | 211°C to 212°C |
Flash Point | 108°C (226°F) |
Quantity | 50 g |
UN Number | UN2430 |
Beilstein | 1363564 |
Solubility Information | Slightly soluble in water,soluble in ethanol and ethyl ether. |
Formula Weight | 136.2 |
Percent Purity | 98% |
Chemical Name or Material | 4-Isopropylphenol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'