missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11429603
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118427-29-5
Molecular Formula C9H14N2
Molecular Weight (g/mol) 150.23
MDL Number MFCD00013461
InChI Key ZYATZFJUOXJFPY-UHFFFAOYSA-N
Synonym 4-isopropylphenylhydrazine hydrochloride, 4-isopropylphenyl hydrazine hydrochloride, 1-4-isopropylphenyl hydrazine hydrochloride, 4-propan-2-yl phenyl hydrazine hydrochloride, hydrazine, 4-1-methylethyl phenyl-, monohydrochloride, 4-isopropylphenylhydrazinehydrochloride, pubchem7533, acmc-1cbwc, 4-propan-2-ylphenyl hydrazine hydrochloride, ksc183e0j
PubChem CID 2733248
IUPAC Name (4-propan-2-ylphenyl)hydrazine;hydrochloride
SMILES CC(C)C1=CC=C(NN)C=C1

Specifications

Melting Point 203°C (decomposition)
Quantity 1 g
Formula Weight 186.69
Percent Purity 98%
Chemical Name or Material 4-Isopropylphenylhydrazine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'