Learn More
4-Keto-4,5,6,7-tetrahydrothianaphthene, 97%, Thermo Scientific Chemicals
£61.90 - £352.00
Chemical Identifiers
CAS | 13414-95-4 |
---|---|
Molecular Formula | C8H8OS |
Molecular Weight (g/mol) | 152.21 |
MDL Number | MFCD00005861 |
InChI Key | GJEKNELSXNSYAQ-UHFFFAOYSA-N |
Synonym | 6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro, 6,7-dihydro-1-benzothiophen-4 5h-one, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 4,5,6,7-tetrahydro-4-benzothiophenone |
PubChem CID | 83418 |
IUPAC Name | 6,7-dihydro-5H-1-benzothiophen-4-one |
SMILES | C1CC2=C(C=CS2)C(=O)C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13414-95-4 | |
152.21 | |
GJEKNELSXNSYAQ-UHFFFAOYSA-N | |
83418 | |
C1CC2=C(C=CS2)C(=O)C1 |
C8H8OS | |
MFCD00005861 | |
6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro, 6,7-dihydro-1-benzothiophen-4 5h-one, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 4,5,6,7-tetrahydro-4-benzothiophenone | |
6,7-dihydro-5H-1-benzothiophen-4-one |
Specifications
13414-95-4 | |
100.0 | |
White to Yellow | |
110°C | |
96% min. (GC) | |
1.5895 to 1.5915 | |
6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro, 6,7-dihydro-1-benzothiophen-4 5h-one, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 4,5,6,7-tetrahydro-4-benzothiophenone | |
C1CC2=C(C=CS2)C(=O)C1 | |
152.21 | |
152.21 | |
Low Melting Solid or Liquid After Melting |
96.0 | |
34°C to 40°C | |
120°C (10.0 mmHg) | |
Authentic | |
C8H8OS | |
MFCD00005861 | |
GJEKNELSXNSYAQ-UHFFFAOYSA-N | |
6,7-dihydro-5H-1-benzothiophen-4-one | |
83418 | |
97% | |
4-Keto-4,5,6,7-tetrahydrothianaphthene |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 236-510-7
- 4-Keto-4,5,6,7-tetrahydrothianaphthene
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only