missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Mercaptophenylacetic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
An useful synthetic intermediate for the preparation of novel anti inflammatory agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
An useful synthetic intermediate for the preparation of novel anti inflammatory agents.
Solubility
Slightly soluble in water. Soluble in Dichloromethane, Ethanol and Methanol
Notes
Air sensitive. Stable under recommended storage conditions. Incompatible with oxidizing agents.
An useful synthetic intermediate for the preparation of novel anti inflammatory agents.
Solubility
Slightly soluble in water. Soluble in Dichloromethane, Ethanol and Methanol
Notes
Air sensitive. Stable under recommended storage conditions. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 105°C to 110°C |
Quantity | 5 g |
Sensitivity | Air sensitive |
Solubility Information | Slightly soluble in water. Soluble in Dichloromethane,Ethanol and Methanol |
Formula Weight | 168.21 |
Percent Purity | 97% |
Chemical Name or Material | 4-Mercaptophenylacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'