missing translation for 'onlineSavingsMsg'
Learn More

4-(Methanesulfonamido)phenylboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 11996871
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380430-57-9
Molecular Formula C7H10BNO4S
Molecular Weight (g/mol) 215.03
MDL Number MFCD02179473
InChI Key NDVJJEADFLTFCD-UHFFFAOYSA-N
Synonym 4-methylsulfonylamino phenylboronic acid, 4-methylsulfonamido phenylboronic acid, 4-methanesulfonylaminophenylboronic acid, 4-methylsulfonylaminophenylboronic acid, 4-methanesulfonylamino phenylboronic acid, 4-methylsulfonamido phenyl boronic acid, 4-methanesulfonamidophenyl boronic acid, 4-methanesulfonamido phenyl boronic acid, 4-methylsulphonyl amino benzeneboronic acid, 4-methylsulfonylamino benzeneboronic acid
PubChem CID 2773537
IUPAC Name [4-(methanesulfonamido)phenyl]boronic acid
SMILES CS(=O)(=O)NC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 234°C to 236°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 215.04
Percent Purity 97%
Chemical Name or Material 4-(Methanesulfonamido)phenylboronic acid
compliance-icons
Product Identifier
  • 4-(Methanesulfonamido)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'