missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-1,2-benzisoxazol-3-amine, 97%, Thermo Scientific™

Product Code. 10591122
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 177995-40-3
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.164
MDL Number MFCD03407354
InChI Key YFNIGSJHKGKXEQ-UHFFFAOYSA-N
Synonym 4-methoxy-1,2-benzisoxazol-3-amine, 4-methoxybenzo d isoxazol-3-amine, 4-methoxy-benzo d isoxazol-3-ylamine, 3-amino-4-methoxy-1,2-benzisoxazole, 1,2-benzisoxazol-3-amine, 4-methoxy, maybridge3_004235, 4-methoxybenzo d isoxazol-3-ylamine, 4-methoxy-1,2-benzisoazole-3-amine, 4-methoxybenzo d isoxazole-3-ylamine, 4-methoxy-1,2-benzisoxazole-3-amine
PubChem CID 2779751
IUPAC Name 4-methoxy-1,2-benzoxazol-3-amine
SMILES COC1=CC=CC2=C1C(=NO2)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material 4-Methoxy-1, 2-benzisoxazol-3-amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.