missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-methylaniline, 98%, Thermo Scientific™

Product Code. 11343726
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-50-1
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00007735
InChI Key CDGNLUSBENXDGG-UHFFFAOYSA-N
Synonym m-cresidine, 2-methyl-4-methoxyaniline, 2-methyl-p-anisidine, 4-methoxy-o-toluidine, benzenamine, 4-methoxy-2-methyl, meta-cresidine, p-anisidine, 2-methyl, 6-amino-3-methoxytoluene, 2-methyl-4-methoxybenzenamine, 4-methoxy-2-methylbenzenamine
PubChem CID 7610
ChEBI CHEBI:82430
IUPAC Name 4-methoxy-2-methylaniline
SMILES CC1=C(C=CC(=C1)OC)N

Specifications

Melting Point 13°C to 14°C
Density 1.065
Boiling Point 248°C to 249°C
Flash Point 124°C (255°F)
Refractive Index 1.5685
Quantity 100 g
UN Number UN2810
Beilstein 774727
Formula Weight 137.18
Percent Purity 98%
Chemical Name or Material 4-Methoxy-2-methylaniline
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'