missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-methylphenylboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 10297597
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 208399-66-0
Molecular Formula C8H11BO3
Molecular Weight (g/mol) 165.98
MDL Number MFCD02684315
InChI Key AMSQNQJCBXQYEX-UHFFFAOYSA-N
Synonym 4-methoxy-2-methylphenyl boronic acid, 2-methyl-4-methoxyphenylboronic acid, 4-methoxy-2-methylbenzeneboronic acid, 4-methoxy-2-methylphenyl boranediol, 4-methoxy-2-methyl-phenyl boronic acid, 4-methoxy-2-methylphenbylboronic acid, 4-methoxy-2-methylphenylboronicacid, boronic acid, 4-methoxy-2-methylphenyl, 2-methyl-4-methoxybenzeneboronic acid, acmc-1cge6
PubChem CID 2773486
IUPAC Name (4-methoxy-2-methylphenyl)boronic acid
SMILES COC1=CC=C(B(O)O)C(C)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 148.0°C to 161.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 165.98
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 4-Methoxy-2-methylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxy-2-methylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'